Tolbutamide, Supelco®

Supplier: SUPELCO

T0891-100G T0891-25G T0891-500G
T0891-100GEA 94.5 EUR
T0891-100G T0891-25G T0891-500G
Tolbutamide, Supelco®
Tolbutamide
Formula: C₁₂H₁₈N₂O₃S
MDL Number: MFCD00027169
CAS Number: 64-77-7
EINECS: 200-594-3
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e7ca1a6260eccf53052f76a0140cbaf7384917b3&vid=e7ca1a6260eccf53052f76a0140cbaf7384917b3&prid=web&clid=SAPDAM https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=160eb2ce71bea4335a79273c4a6a2c6299ac0bdf&vid=160eb2ce71bea4335a79273c4a6a2c6299ac0bdf&prid=web&clid=SAPDAM https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a633ad324da39d14a7607e8a0e1b21d731b0a440&vid=a633ad324da39d14a7607e8a0e1b21d731b0a440&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR