Stand-up lectern tables, 304 stainless steel
Supplier: TEKNOMEK
DELT-1SHMS DELT-1SHS DELT-2SHMS DELT-2SHS DELT-FL DELT-FX DELT-MS DELT-S
TEKMDELT-1SHMSEA 0 EUR
TEKMDELT-1SHMS TEKMDELT-1SHS TEKMDELT-2SHMS TEKMDELT-2SHS TEKMDELT-FL TEKMDELT-FX TEKMDELT-MS TEKMDELT-S
Stand-up lectern tables, 304 stainless steel
Furniture Tables and Workbenches
304 grade stainless steel lecterns with sloped top that are perfect for standing at. Durable and strong, with hygienic design features to minimise risk. They are fully-welded, with reduce dirt traps to reduce risk. Choose from freestanding, wall mounted or mobile, with a wide choice of shelf options too.
  • 304-grade EU sourced stainless steel
  • Supplied fully welded, no construction or assembly required
  • Ideal for standing
  • Inclined top with lip to hold documents in place
  • Hygienic design throughout
  • Brushed dull polish finish
  • Choice of mobile or static
  • Choice of
More Product Information