Lift lid PPE dispensers
Supplier: TEKNOMEK
HWDP-L1V25X30 HWDP-L2V50X30 HWDP-L3V75X30 HWDP-L4V10X30 HWDP-L1N25X30 HWDP-L2N50X30 HWDP-L3N75X30 HWDP-L4N10X30 HWDPL1V25X30PE HWDPL2V50X30PE HWDPL3V75X30PE HWDPL4V10X30PE HWDPL1N25X30PE HWDPL2N50X30PE HWDPL3N75X30PE HWDPL4N10X30PE
TEKMHWDP-L1V25X30EA 0 EUR
TEKMHWDP-L1V25X30 TEKMHWDP-L2V50X30 TEKMHWDP-L3V75X30 TEKMHWDP-L4V10X30 TEKMHWDP-L1N25X30 TEKMHWDP-L2N50X30 TEKMHWDP-L3N75X30 TEKMHWDP-L4N10X30 TEKMHWDPL1V25X30PE TEKMHWDPL2V50X30PE TEKMHWDPL3V75X30PE TEKMHWDPL4V10X30PE TEKMHWDPL1N25X30PE TEKMHWDPL2N50X30PE TEKMHWDPL3N75X30PE TEKMHWDPL4N10X30PE
Lift lid PPE dispensers
PPE Dispensers
Our lift lid PPE dispenser comes in a variety of sizes to suit most applications.
  • Brushed dull polish finish
  • No plastic parts
  • Mounts proud of wall or pedestal for easy cleaning
  • Pedestal fixings included
  • Optional vision slots
The Lift lid PPE dispenser is suitable for various items of PPE including beard snoods, hair nets, face masks, ear plugs and shoe covers. Lifting tabs on the lid for ease of opening and filling, they double as a locking point by way of a padlock (padlock not supplied). Hygienic backwards sloping top and drainage slots for rapidMore Product Information