Cleanroom tables, hygienic design, 304 grade stainless steel
Supplier: TEKNOMEK
TATACTNUMS1260 TATACTNUMS1560 TATACTNUMS1860 TATACTNUMS6060 TATACTNUMS9060 TATACTNUS12X60 TATACTNUS18X60 TATACTNUS60X60 TATACTNUS90X60 TATARTNUMS1260 TATARTNUMS1560 TATARTNUMS6060 TATARTNUMS9060 TATARTNUS12X60 TATARTNUS15X60 TATARTNUS18X60 TATARTNUS60X60 TATARTNUS90X60 TATARTUPMS1260 TATARTUPMS1560 TATARTUPMS1860 TATARTUPMS6060 TATARTUPMS9060 TATARTUPS12X60 TATARTUPS15X60 TATARTUPS18X60 TATARTUPS90X60 TATAUSNUMB9060 TATAUSNUMS1260 TATAUSNUMS1560 TATAUSNUMS1860 TATAUSNUMS6060 TATAUSNUMS9060 TATAUSNUS12X60 TATAUSNUS15X60 TATAUSNUS18X60 TATAUSNUS60X60 TATAUSNUS90X60 TATAUSUPMS1260 TATAUSUPMS1560 TATAUSUPMS1860 TATAUSUPMS9060 TATAUSUPS12X60 TATAUSUPS15X60 TATAUSUPS18X60 TATAUSUPS60X60 TATAUSUPS90X60
TEKMTATACTNUMS1260EA 0 EUR
TEKMTATACTNUMS1260 TEKMTATACTNUMS1560 TEKMTATACTNUMS1860 TEKMTATACTNUMS6060 TEKMTATACTNUMS9060 TEKMTATACTNUS12X60 TEKMTATACTNUS18X60 TEKMTATACTNUS60X60 TEKMTATACTNUS90X60 TEKMTATARTNUMS1260 TEKMTATARTNUMS1560 TEKMTATARTNUMS6060 TEKMTATARTNUMS9060 TEKMTATARTNUS12X60 TEKMTATARTNUS15X60 TEKMTATARTNUS18X60 TEKMTATARTNUS60X60 TEKMTATARTNUS90X60 TEKMTATARTUPMS1260 TEKMTATARTUPMS1560 TEKMTATARTUPMS1860 TEKMTATARTUPMS6060 TEKMTATARTUPMS9060 TEKMTATARTUPS12X60 TEKMTATARTUPS15X60 TEKMTATARTUPS18X60 TEKMTATARTUPS90X60 TEKMTATAUSNUMB9060 TEKMTATAUSNUMS1260 TEKMTATAUSNUMS1560 TEKMTATAUSNUMS1860 TEKMTATAUSNUMS6060 TEKMTATAUSNUMS9060 TEKMTATAUSNUS12X60 TEKMTATAUSNUS15X60 TEKMTATAUSNUS18X60 TEKMTATAUSNUS60X60 TEKMTATAUSNUS90X60 TEKMTATAUSUPMS1260 TEKMTATAUSUPMS1560 TEKMTATAUSUPMS1860 TEKMTATAUSUPMS9060 TEKMTATAUSUPS12X60 TEKMTATAUSUPS15X60 TEKMTATAUSUPS18X60 TEKMTATAUSUPS60X60 TEKMTATAUSUPS90X60
Cleanroom tables, hygienic design, 304 grade stainless steel
Furniture Tables and Workbenches
Fully welded stainless steel tables are a very versatile work surface, intelligently designed to remove potential trap points. Features include diamond braces - at 300 mm from the ground they aid ease of cleaning. Choose between diamond centre tie or rear tie bar or undershelf to suit your needs.
  • 304 grade stainless steel
  • Supplied fully welded, no construction or assembly
  • Brushed dull polish finish
  • 1.5 mm thick stainless steel sheet
  • Choice of static or mobile; with undershelf option
  • Easy access diamond tie bar for efficient cleaning
  • Space for extra storage
More Product Information