Autoclaves, Prestige Medical Classic Models
Supplier: TUTTNAUER
210048 210052 210001
TUTT210048EA 1640 EUR
TUTT210048 TUTT210052 TUTT210001
Autoclaves, Prestige Medical Classic Models
Sterilisers Autoclaves (Steam Sterilisers)
The Classic is reliable and trusted worldwide, designed to give you the most cost effective sterilisation solution. This light, compact, portable and robust top loading autoclave is simple to use, fully automatic with easy, one touch operation and guaranteed to meet your needs.
  • Simple, one-step operation to start the cycle
  • Two chamber sizes available, standard (9 L) or extended (12 L)
  • Supplied with all standard accessories, including basket
  • Maximum instrument length is 228 mm for 9 L models, and 268 mm for the 12 L units
  • Minimum sterilisation time is 28
More Product Information