Centrifugal filter units, Amicon® Ultra-15
Supplier: Merck
UFC901008 UFC901024 UFC901096 UFC903008 UFC903024 UFC903096 UFC910008 UFC910024 UFC910096 UFC905008 UFC905024 UFC900308 UFC900324 UFC905096 UFC900396
UFC901008EA 152 EUR
UFC901008 UFC901024 UFC901096 UFC903008 UFC903024 UFC903096 UFC910008 UFC910024 UFC910096 UFC905008 UFC905024 MILFUFC900308 MILFUFC900324 MILFUFC905096 MILFUFC900396
Centrifugal filter units, Amicon® Ultra-15
Filters Centrifugal Filters
These centrifugal filter units can be used for a number of applications: concentration of biological samples containing antigens, antibodies, enzymes, nucleic acids, or microorganisms; purification of macromolecular components found in tissue culture extracts or cell lysates and protein removal prior to HPLS; concentration of dilute or pre-purified proteins from column eluents; desalting, buffer exchange and protein dialysis; size exclusion and protein fractionation.
  • High retentate recovery of >90%; 100% integrity tested
  • Vertical membrane reduces
More Product Information