Anti-SYT1 Mouse Monoclonal Antibody [clone: 13B752]

Supplier: US Biological
030674
USBI030674EA 626 EUR
USBI030674
Anti-SYT1 Mouse Monoclonal Antibody [clone: 13B752]
Antibodies
Type: Primary
Antigen: SYT1 (synaptotagmin I)
Clonality: Monoclonal
Clone: 13B752
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR