Anti-PRR24 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Supplier: US Biological
040505-PE
USBI040505-PEEA 884 EUR
USBI040505-PE
Anti-PRR24 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antibodies
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: PRR24 (proline rich 24)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol PRR24
Environmentally Preferable
Antigen name proline rich 24
Antibody type Primary
Clonality Polyclonal
Conjugation PE (Phycoerythrin)
Reactivity Human
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
Epitope C-terminal
Form liquid
Purification purified by Protein A chromatography
Storage buffer PBS, pH 7,2
Storage temperature Cool
Shipping temperature Ice
Tested applications Fluoresence ELISA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR