Anti-UPP1 Mouse Monoclonal Antibody [clone: 2F5]

Supplier: US Biological
135121
USBI135121EA 655 EUR
USBI135121
Anti-UPP1 Mouse Monoclonal Antibody [clone: 2F5]
Antibodies
Recommended dilution: Immunochemistry: 3?g/ml

Type: Primary
Antigen: UPP1 (uridine phosphorylase 1)
Clonality: monoclonal
Clone: 2F5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR