Anti-OGFOD1 Rabbit Polyclonal Antibody

Supplier: US Biological
146801
USBI146801EA 647 EUR
USBI146801
Anti-OGFOD1 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: Western Blot: 1 ?g/ml

Type: Primary
Antigen: OGFOD1 (2-oxoglutarate and iron-dependent oxygenase domain containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR