Anti-BAK1 Rabbit monoclonal antibody unconjugated [clone: D2D3]

Supplier: US Biological
B0055-10J
USBIB0055-10JEA 674 EUR
USBIB0055-10J
Anti-BAK1 Rabbit monoclonal antibody unconjugated [clone: D2D3]
Antibodies
Recommended dilution: Western Blot: 1:1000; Immunoprecipitation: 1:100; Immunochemistry: 1:800

Type: Primary
Antigen: BAK1 (BCL2-antagonist/killer 1)
Clonality: monoclonal
Clone: D2D3
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR