Anti-Bone Morphogenic Protein Receptor 1A Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Supplier: US Biological
B2553-60B-APC
USBIB2553-60B-APCEA 884 EUR
USBIB2553-60B-APC
Anti-Bone Morphogenic Protein Receptor 1A Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antibodies
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: Bone Morphogenic Protein Receptor 1A
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR