Anti-CLOCK Rabbit Polyclonal Antibody

Supplier: US Biological
C5843-01-100
USBIC5843-01-100EA 742 EUR
USBIC5843-01-100
Anti-CLOCK Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CLOCK (clock circadian regulator)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Drosophila
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR