Anti-IL36B Rabbit Polyclonal Antibody (Biotin)

Supplier: US Biological
I7663-24W-BIOT
USBII7663-24W-BIOTEA 884 EUR
USBII7663-24W-BIOT
Anti-IL36B Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: IL36B (interleukin 36, beta)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR