Anti-MAP Kinase p44/42 Rabbit Polyclonal Antibody

Supplier: US Biological
M2352-20L
USBIM2352-20LEA 379 EUR
USBIM2352-20L
Anti-MAP Kinase p44/42 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: Western Blot:1:500-1:1000

Type: Primary
Antigen: MAP Kinase p44/42
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR