Anti-MDM2 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Supplier: US Biological
M2800-04C-FITC
USBIM2800-04C-FITCEA 884 EUR
USBIM2800-04C-FITC
Anti-MDM2 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antibodies
Recommended dilution: FLISA: 1:1000; Western Blot: 1:50-1:100; Immunochemistry: 1:10-1:50
Do not freeze!

Type: Primary
Antigen: MDM2 (MDM2 oncogene, E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR