Anti-MDR Antibody Kit, BioAssay™ Mouse Monoclonal Antibody [clone: JSB-1, LRP-56, MRPm6, MRPr1]

Supplier: US Biological
M2825-02
USBIM2825-02EA 1300 EUR
USBIM2825-02
Anti-MDR Antibody Kit, BioAssay™ Mouse Monoclonal Antibody [clone: JSB-1, LRP-56, MRPm6, MRPr1]
Antibodies
Type: Primary
Antigen: MDR Antibody Kit, BioAssay™
Clonality: monoclonal
Clone: JSB-1, LRP-56, MRPm6, MRPr1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1+IgG2b+IgG1+IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR