Anti-Mycoplasma pneumoniae Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: US Biological
M9750-19C
USBIM9750-19CEA 636 EUR
USBIM9750-19C
Anti-Mycoplasma pneumoniae Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Recommended dilution: Immunochemistry: 1:20-1:200; ELISA: 1:200-1:1000; Western Blot: 1:20-1:200

Type: Primary
Antigen: Mycoplasma pneumoniae
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR