Anti-Mycoplasma pneumoniae Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: US Biological
M9750-27A
USBIM9750-27AEA 512 EUR
USBIM9750-27A
Anti-Mycoplasma pneumoniae Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Recommended dilution: ELISA: 1:1200-1:1000; Immunochemistry: 1:20-1:200; Western Blot: 1:20-1:200; Immunofluorescence: 1:1000

Type: Primary
Antigen: Mycoplasma pneumoniae
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR