Anti-ROR2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: US Biological
R2999-61C-HRP
USBIR2999-61C-HRPEA 884 EUR
USBIR2999-61C-HRP
Anti-ROR2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 1:500; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: ROR2 (receptor tyrosine kinase-like orphan receptor 2)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR