Anti-TOPORS Mouse Monoclonal Antibody [clone: 5G11]

Supplier: US Biological
T1001-25A
USBIT1001-25AEA 703 EUR
USBIT1001-25A
Anti-TOPORS Mouse Monoclonal Antibody [clone: 5G11]
Antibodies
Recommended dilution: Immunofluorescence: 10 ?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: TOPORS (topoisomerase I binding, arginine/serine-rich, E3 ubiquitin protein ligase)
Clonality: monoclonal
Clone: 5G11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR