Anti-UPP1 Chicken Polyclonal Antibody

Supplier: US Biological
U2283-01
USBIU2283-01EA 474 EUR
USBIU2283-01
Anti-UPP1 Chicken Polyclonal Antibody
Antibodies
Recommended dilution: Western Blot: 1:1000-1:2000

Type: Primary
Antigen: UPP1 (uridine phosphorylase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR