Construction and maintenance cleanroom tapes
Supplier: ULTRATAPE
1160BL200-P3G
UTAP1160BL200-P3GEA 150 EUR
UTAP1160BL200-P3G
Construction and maintenance cleanroom tapes
Tapes
These cleanroom tapes are made of vinyl with rubber adhesive and are suitable for construction and maintenance.
  • Combines particle-free cleanliness with the ability to adhere to a variety of materials, including plastic and metal
  • Abrasion and moisture resistant and unaffected by most chemicals
  • Performs well over a wide temperature range
  • Can be written on by cleanroom pen
Suitable for use in ISO Class 4 environments.
Length: 32,9 m
Different colours available.
Packaging: Supplied double-bagged.