Uvex Silv-Air Premium Respirators, FFP 2
Supplier: UVEX
8735321 8735311 8735211 8735310 8735101 8735111 8735200 8735221 8735210 8735110 8735201 8735100
UVEX8735321EA 151 EUR
UVEX8735321 UVEX8735311 UVEX8735211 UVEX8735310 UVEX8735101 UVEX8735111 UVEX8735200 UVEX8735221 UVEX8735210 UVEX8735110 UVEX8735201 UVEX8735100
Uvex Silv-Air Premium Respirators, FFP 2
Respirators Disposable Respirators Particulate Respirators
The Uvex silv-Air premium product line comprises classic, white folding respirators that adapt to different face shapes. The headband and integrated sealing lip in all protection classes mean the masks are easy to put on correctly as well as making them comfortable to wear, even for extended periods.
  • Particle-filtering folding mask for small to medium and larger face shapes
  • White colour
  • Compatible with Uvex safety eyewear
  • Adjustable head band offering easy handling and comfortable fit
  • Wide, seamless headband for ensuring a comfortable and secure fit
More Product Information