Bump caps, uvex u-cap sport
Supplier: UVEX
9794.401 9794.402 9794.400 9794.403 9794.404 9794.407 9794.408 9794.409
111-2441EA 34.6 EUR
111-2441 UVEX9794.402 UVEX9794.400 UVEX9794.403 UVEX9794.404 UVEX9794.407 UVEX9794.408 UVEX9794.409
Bump caps, uvex u-cap sport
Head Protection Bump Caps
These caps feature an innovative hard shell element for reliable impact protection.
  • Protect against abrasions and impact injuries
  • Hard shell element in 'armadillo' design with honeycomb structure and shock-absorbing elements
  • Integrated reflective piping on the brim and textile components
  • Can be quickly and easily fitted to the head using the adjustable Velcro® fastening
  • Available with short peak for a wider field of upward vision or with long peak for increased sun protection
The armadillo design provides additional flexibility and ergonomicallyMore Product Information