Vacuum manifolds, Vac Elut™ 20
Supplier: VARIAN
12234100 12234101 12234105 12234102 12234103
551-0394EA 1990 EUR
551-0394 VARI12234101 VARI12234105 VARI12234102 VARI12234103
Vacuum manifolds, Vac Elut™ 20
Manifolds
The Vac Elut™ 20 vacuum control valve, vacuum gauge, and quick release valve are mounted on the lid, away from the corrosive waste stream and within convenient reach. The solvent-resistant PP rack is available in a variety of sizes to accommodate collection tubes commonly used in sample preparation.
Delivery information: Manifold sets include the glass basin, lid cover, collection rack and vacuum gauge assembly.