Solid phase extraction cartridges, Bond Elut™ SAX
Supplier: VARIAN
12162044B 12166013B 12256013 12113017 12113043 12102044 12102017 12102079 12256051 12102144 12102125 12102087 12256021 12256029 12256037 14113043 14256013 14256021 14256029 14102079 14102017 14102044 7550801C 75508025 75508050 75408050 12102017T 12102017TB 12102044T
551-0379EA 431 EUR
551-0379 551-0385 549-0823 VARI12113017 551-0348 549-0780 551-0311 549-0793 549-0832 549-0805 549-0804 549-0794 549-0824 549-0826 549-0828 VARI14113043 549-0859 549-0864 549-0868 549-0854 549-0843 549-0848 VARI7550801C VARI75508025 549-0875 549-0874 549-0777 549-0778 549-0781
Solid phase extraction cartridges, Bond Elut™ SAX
Chromatography Columns
Bond Elut™ SAX is a strong anion exchange sorbent ideally suited for the extraction of compounds such as carboxylic acids, which may not retain effectively on weak anion exchange sorbents. The sorbent is supplied in the chloride counter ion form. You can modify the selectivity of the phase by replacing the chloride ion with other anions, such as acetate and hydroxide.
  • Retains compounds that elute from weak anion exchange sorbents
  • Selectivity can be user-modified for increased flexibility
  • Minimal non-polar interactions