Tubing kit for low volatile organic solvents
Supplier: VARIAN
9910124200 9910124300 9910124400 9910124500
VARI9910124200EA 900 EUR
VARI9910124200 VARI9910124300 VARI9910124400 VARI9910124500
Tubing kit for low volatile organic solvents
Tubing Chromatography Tubing
The ICP-OES tubing kit for low volatile organic solvents includes:
  • Two packs PVC Solva-flex peristaltic pump tubing, 12/pk (one each black/black and white/white)
  • One FEP transfer tube (spray chamber to torch)
  • Drain tubing, 1 m
  • Tubing for waste line from spray chamber, 1 m
  • One pack EzyFit sample inlet connectors, 10/pk
  • Nebuliser capillary tubing, 2 m
  • Barb connectors for joining tubing (5 each 1/16 to 1/8 in Int.Ø. and 5 each 1/16 to 1/16 in Int.Ø.)