Bottles, narrow neck, round, LDPE, with screw caps
Supplier: VITLAB
138193
VITL138193EA 112 EUR
VITL138193
Bottles, narrow neck, round, LDPE, with screw caps
Bottles Plastic Bottles
LDPE, translucent.
  • Leakproof, robust and versatile
  • Squeezable, so these bottles make excellent containers for dropping or squirting
  • Good reset properties
Excellent for sample collection, storing and transporting liquids.