VWR® Talon®, Clamps, three pronged
Supplier: VWR Collection
21570-200
VWRU21570-200EA 44.5 EUR
VWRU21570-200
VWR® Talon®, Clamps, three pronged
Clamps General Purpose Clamps
Zinc, nickel plated. These clamps securely hold every type of glassware and apparatus.
  • Large grip adjustment range
  • Dual adjustment
  • Nickel-plated zinc offers high tensile strength
  • Long, seamless tubing attaches clamp head securely and offers easy positioning in the deepest fume hoods
  • Supplied with non slip vinyl sleeves, and fibreglass covers for temperatures above 100 °C
The clamps are constructed with round extension arms that allow the clamps to be rotated 360°. Extension arms allow placement of apparatus at various distances from lab frames withoutMore Product Information