Dropping bottles with stoppers
Supplier: WITEG LABORTECHNIK
5.883.100
WITG5.883.100EA 10.8 EUR
WITG5.883.100
Dropping bottles with stoppers
Bottles Dropping Bottles
Glass, amber.
  • Good chemical resistance
  • Amber coloured to protect light sensitive substances
Dropping bottle with grooved flat stopper.