Digital orbital shakers, WiseShake® SHO-2D
Supplier: WITEG LABORTECHNIK
DH.WSO04012
WITGDH.WSO04012EA 1330 EUR
WITGDH.WSO04012
Digital orbital shakers, WiseShake® SHO-2D
Shakers Orbital Shakers
Digital orbital shaker with programmable feedback control. Unit has a backlit digital LCD display, is ideal for culturing cells to hybridisation, staining and destaining gels, and combinatorial chemistry.
  • Optimal platform rotation for intensive mixing
  • Brushless DC motor (low noise and long life cycle)
  • With Jog-Shuttle control system
  • High precision digital rotation speed control
  • With Programmable operation mode, storage function, alarm, over-load protection and locking mode for experimental safety
  • Smooth start and change of shaking speed
  • Timer: 99 h 59 min
More Product Information