Masterflex® Microbore Transfer Tubing, Tygon® ND-100-80, 0.020" ID x 0.060" OD; 100 Ft
Supplier: Avantor Fluid Handling
Clear formulation for easier sample viewing.
- Ideal for general-purpose, single-use, and extended fluid path applications
- Single-use custom lengths and configurations available to minimise downtime
- Tubing size: 0.020" ID x 0.060" OD
<B>Formulation:</B> Tygon ND-100-80<br /><B>Applications:</B> General laboratory and pharmaceutical use, especially with precision injection or dispensing applications. Also excellent with low-flow peristaltic pump applications.<br /><B>Characteristics:</B> Extremely flexible. Non-toxic; nonpyrogenic; biocompatible. Crystal clear.<br /><B>Certifications:</B> Resin meets USP Class VI requirements.<br /><B>Temperature range:</B> -25 to 185 °F (-31 to 85 °C)<br /><B>Sterilize:</B> Autoclave, ethylene oxide, or gamma irradiation.