Masterflex® Microbore Transfer Tubing, Tygon® ND-100-80, 0.050" ID x 0.090" OD; 100 Ft
Supplier: Avantor Fluid Handling
VWR Item#MFLX06419-05
Your Organization's Price:
Retrieving
Clear formulation for easier sample viewing.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Single-use custom lengths and configurations available to minimise downtime
  • Tubing size: 0.050" ID x 0.090" OD
<B>Formulation:</B> Tygon ND-100-80<br /><B>Applications:</B> General laboratory and pharmaceutical use, especially with precision injection or dispensing applications. Also excellent with low-flow peristaltic pump applications.<br /><B>Characteristics:</B> Extremely flexible. Non-toxic; nonpyrogenic; biocompatible. Crystal clear.<br /><B>Certifications:</B> Resin meets USP Class VI requirements.<br /><B>Temperature range:</B> -25 to 185 °F (-31 to 85 °C)<br /><B>Sterilize:</B> Autoclave, ethylene oxide, or gamma irradiation.