Masterflex® Remote Control Cable Connectors, Avantor®
Supplier: Avantor Fluid Handling
Create your own cable for optimization of your Masterflex® drive.
- Choose the correct connection for your Masterflex® pump
Combine these connectors with 20-22 gauge shielded wire to create your own interface cable. Maximum cable length is 1000 ft (300 m).