Masterflex® Gold-colored IDentifier Hinge Clamp, 1/2-3/4" Clamp Size, 304 SS; 1/EA
Supplier: Avantor Fluid Handling
VWR Item#MFLX30562-51
Your Organization's Price:
Retrieving
Easy visual identification of critical joints for review in your process.
  • Constructed of gold-coloured 304 Stainless Steel
Easily distinguishable from standard stainless steel clamps from a distance, these gold-coloured sanitary clamps are identical in size and function and can be used interchangeably with standard stainless steel clamps. Clamps feature a TiN (titanium nitride) coating. Clamps are available in a variety of sizes.