GF Signet Rotor-X High Flow Sensor for 1/2 to 4" Pipe, PVDF Body, Hastelly C Shaft
Supplier: Avantor Fluid Handling
VWR Item#MFLX05618-13
Your Organization's Price:
Retrieving
Open-core design ensures accurate, linear output to ±1%.
  • Eliminate cavitation and minimize pressure drop
  • Flow velocity of 1 to 20 ft/sec (0.3 to 6 m/sec)
Sensors contain an open-core rotor with a small magnet in each of the four blades. As they rotate, the magnets pass a coil in the sensor. The transducer generates a linear frequency output that is proportional to the flow velocity. This signal travels up to 200 feet without amplification.<br /><br />Open-core design eliminates cavitation at flow velocities from 1 to 20 ft/second. Closed or solid paddle-wheel designs create air bubbles at these velocities, causing a nonlinear, nonrepeatable signal.<br /><br />Sensor materials resist most chemicals. All models have PVDF rotor and VITON® O-ring; and include a 25-ft connecting cable.<br /><br />ROTOR-X flow sensors are compatible with all of our line-, battery-, and sensor-powered flowmeters, accumulators, and controllers. Mount in any type of pipe using an installation fitting—order separately.
<b>Delivery information: </b>Includes 25-ft connecting cable.