Masterflex® Plastic Ball Valves, Two-way Right Angle, Avantor®
Supplier: Avantor Fluid Handling
VWR Item#MFLX01378-52
Your Organization's Price:
Retrieving
Nonmetallic wetted parts ideal for water and mild aqueous solutions.
  • Maximum pressure: 125 psi (8.6 bar)
  • Maximum temperature: 140 °F (60 °C)
Ball valves are designed primarily for on/off service.