Masterflex® Ball valve, 2-way right angled, BUNA, 1/4" NPT(M) x 1/4" compression
Supplier: Avantor Fluid Handling
Nonmetallic wetted parts ideal for water and mild aqueous solutions.
- Maximum pressure: 125 psi (8.6 bar)
- Maximum temperature: 140 °F (60 °C)
Ball valves are designed primarily for on/off service.