Masterflex® Ball valve, 2-way right angled, BUNA, 1/4" NPT(F) x 1/4" John Guest
Supplier: Avantor Fluid Handling
VWR Item#MFLX01379-17
Your Organization's Price:
Retrieving
Nonmetallic wetted parts ideal for water and mild aqueous solutions.
  • Maximum pressure: 125 psi (8.6 bar)
  • Maximum temperature: 140 °F (60 °C)
Ball valves are designed primarily for on/off service.