SANI-TECH Ultra C Ultra-Pure Platinum-Cured Silicone Tubing, 1/8" ID x 1/4" OD; 100 Ft
Supplier: Avantor Fluid Handling
VWR Item#MFLX96201-63
Your Organization's Price:
Retrieving
Don't let extractables ruin your formulations.
  • Exceptionally low TOCs minimize fluid contamination
  • Ultra-pure silicone will not impart flavors or odors
  • Superior pumping life reduces downtime for tubing changes
Applications: Sterile fill and transfer, cell harvest, media processing, sanitary peristaltic pumping, production filtration and fermentation, bioreactor process lines, and pharmaceutical delivery.<br />Characteristics: Excellent flexibility, long pump life, non-yellowing, tasteless and odorless, nontoxic, translucent.<br />Certifications: Elastomer is FDA-compliant and meets USP Class VI, EP 3.1.9, and ISO 10993 biocompatibility criteria. Tubing is manufactured and packaged in a certified cleanroom.<br />Temperature range: -80 to 500 °F (-62 to 260 °C).<br />Sterilize: Autoclave, ethylene oxide, or gamma irradiation (up to 5.0 Mrad).