2-Mercaptothiazole ≥97%

Supplier: ALFA AESAR

L07058.03 L07058.06 L07058.14
L07058.03EA 46.2 EUR
L07058.03 L07058.06 L07058.14
2-Mercaptothiazole ≥97%
2-Mercaptothiazole

Purity: 97% CAS No.: 82358-09-6 Molecular Formula: C3H3NS2 Formula Weight: 117.19 Melting Point: 77-80° Beilstein Reference: 635950

Formula: C₃H₃NS₂
MDL Number: MFCD00022449
CAS Number: 82358-09-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR