2-(4-Methoxyphenyl)-4-thiazolecarboxylic acid ≥97%
Supplier: ALFA AESAR
|
Warning
|
H54606.MD
H54606.03
H54606.06
H54606.MDEA
0
EUR
H54606.MD
H54606.03
H54606.06
2-(4-Methoxyphenyl)-4-thiazolecarboxylic acid ≥97%
2-(4-Methoxyphenyl)-4-thiazolecarboxylic acid
Purity: 97%
CAS No.: 57677-80-2
Molecular Formula: C11H9NO3S
Formula Weight: 235.26
MDL No.: MFCD03791177
Formula:
C₁₁H₉NO₃S Storage Temperature: Ambient |
MDL Number:
MFCD03791177 CAS Number: 57677-80-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...