Zinc nitride ≥99% (metals basis)

Supplier: ALFA AESAR
Warning

88212.04 88212.09
88212.04EA 0 EUR
88212.04 88212.09
Zinc nitride ≥99% (metals basis)
Zinc nitride

Purity: 99% CAS No.: 1313-49-1 Molecular Formula: Zn3N2 Formula Weight: 224.12 Physical Form: Powder Density: 6.22 MDL No.: MFCD00061327

Formula: N₂Zn₃
MW: 224.12 g/mol
Density: 6.22
Storage Temperature: Ambient
MDL Number: MFCD00061327
CAS Number: 1313-49-1
EINECS: 215-207-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR