Iron silicide ≥99.9% (metals basis)
Supplier: ALFA AESAR
|
Warning
|
14019.14
14019.22
14019.14EA
161
EUR
14019.14
14019.22
Iron silicide ≥99.9% (metals basis)
Iron silicide
Purity: 99.9%
CAS No.: 12022-95-6
Molecular Formula: FeSi
Formula Weight: 83.93
Physical Form: -20 Mesh
Odour: Odourless
MDL No.: MFCD00016086
Formula:
FeSi Melting Pt: 1410 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016086 CAS Number: 12022-95-6 EINECS: 234-671-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...