N,N-Dimethylformamide dibenzyl acetal tech. 80%, Technical Grade
|
Warning
|
A11281.09
A11281.18
A11281.30
A11281.09EA
110
EUR
A11281.09
A11281.18
A11281.30
N,N-Dimethylformamide dibenzyl acetal tech. 80%, Technical Grade
N,N-Dimethylformamide dibenzyl acetal
Purity: 85%
CAS No.: 2016-04-8
Molecular Formula: (CH3)2NCH(OCH2C6H5)2
Formula Weight: 271.36
Boiling Point: 138-140°/0.5mm
MDL No.: MFCD00014869
Formula:
C₁₇H₂₁NO₂ MW: 271.36 g/mol Boiling Pt: 138…140 °C (0.5 mmHg) Density: 1.045 Storage Temperature: Ambient |
MDL Number:
MFCD00014869 CAS Number: 2016-04-8 EINECS: 217-946-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...