3-Chloro-p-toluidine hydrochloride ≥97%
Supplier: ALFA AESAR
|
Warning
|
H31342.36
H31342.06
H31342.14
H31342.36EA
0
EUR
H31342.36
H31342.06
H31342.14
3-Chloro-p-toluidine hydrochloride ≥97%
3-Chloro-p-toluidine hydrochloride
Purity: 97%
CAS No.: 7745-89-3
Molecular Formula: C7H8ClN.HCl
Formula Weight: 178.06
Melting Point: ca 260° subl.
MDL No.: MFCD00242925
Formula:
C₇H₉Cl₂N Melting Pt: ∼ 260 °C (subl.) Storage Temperature: Ambient |
MDL Number:
MFCD00242925 CAS Number: 7745-89-3 EINECS: 202-446-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...