N,N-Diethylnicotinamide 97%

Supplier: ACROS ORGANICS
Danger

114471000 114470250
ACRO114471000EA 132 EUR
ACRO114471000 ACRO114470250
N,N-Diethylnicotinamide 97%
N,N-Diethylnicotinamide
Formula: C₁₀H₁₄N₂O
Boiling Pt: 296…300 °C (760 mmHg)
Density: 1.06 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00006386
CAS Number: 59-26-7
EINECS: 200-418-5
UN: 2810
ADR: 6.1 II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5240 to 1.5260 (20°C, 589 nm)
GC ≥99.0 %
Specific optical rotation +39° to +41° (20°C, 589 nm) (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR