4-Fluorphenylmagnesium bromide 0.8 mol/l THF, AcroSeal®
Supplier: ACROS ORGANICS
|
Danger
|
377231000
377238000
377231000.EA
86.4
EUR
377231000.
377238000.
4-Fluorphenylmagnesium bromide 0.8 mol/l THF, AcroSeal®
4-Fluorphenylmagnesium bromide
Formula:
C₆H₄BrFMg Melting Pt: 5 °C Density: 1.01 g/cm³ Flash Pt: –21 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009667 CAS Number: 352-13-6 UN: 3399 ADR: 4.3 II |
Specification Test Results
Appearance | Clear to hazy yellow to dark brown Solution (May develop some turbidity or precipitate) |
Titration Grignard | 14 to 17 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...