Diethyl ether stabilised USP for anesthesia, J.T.Baker®

Danger


Stabilization: Stabilised with BHT (2,6-Di-tert-butyl-p-cresol, 2,6-Di-tert-butyl-4-methylphenol, Ionol) and ethanol

9249-22EA 145.51 EUR
9249-22 9249-02
Diethyl ether stabilised USP for anesthesia, J.T.Baker®
Diethyl ether
Formula: (CH₃CH₂)₂O
MW: 74.12 g/mol
Boiling Pt: 35
Melting Pt: –123 °C
Density: 0.7135 g/cm³ (20 °C)
Flash Pt: –40 °C
Storage Temperature: Ambient
MDL Number: MFCD00011646
CAS Number: 60-29-7
EINECS: 200-467-2
UN: 1155
ADR: 3 I
REACH: 01-2119535785-29
Merck Index: 12,03852
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16f2debd1f18f9bff9952c4a6e5d599a67f94e61&vid=e25ba31ba5c9afb69382c3ca3d66e401980e4bba&prid=web&clid=SAPDAM

Order Now

Specification Test Results

GMP Manufactured Product 0.713 - 0.716
Meets U.S.P Requirements Passes Test
CAUTION: For Manufacturing, processing or repackaging = 0.003 %
Bulk Pharmaceutical Chemical 0.713 - 0.716
USP - Specific Gravity at 25°/25°C 0.713 - 0.716
USP - Acidity Passes Test
USP - Nonvolatile Residue ≤0.003 %
USP - Aldehydes Passes Test
USP - Peroxide (as H₂O₂) ≤0.3 ppm
USP - Low Boiling Hydrocarbons ≤0.2 %
USP - Preservative (C₂H₅OH), For Information Only = 0.2 %
USP - Preservative (H₂O), (w/w) ≤0.2 %
USP - Preservative (BHT) ≥7 ppm
Only the Class 3 solvent Ethanol is likely to be present as a preservative. The actual concentration is reported.
Keep in partly filled, tightly closed, light resistant containers, at a temperature not exceeding 30°C (86°F), remote from fire


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR